Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FYV7
Entrez Gene ID
850757
Gene Name
Gene Aliases
YLR068W
Chromosome Location
Chr.XII: 271008 - 271463 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181955.1 NP_013169.1 1 - 1 345 91

View Full Product Details