Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SKG3
Entrez Gene ID
850884
Gene Name
Gene Aliases
YLR187W
Chromosome Location
Chr.XII: 524865 - 527945 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182074.1 NP_013288.1 1 - 1 2948 106

View Full Product Details