Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STE11
Entrez Gene ID
851076
Gene Name
Gene Aliases
YLR362W
Chromosome Location
Chr.XII: 849866 - 852019 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182251.1 NP_013466.1 1 - 1 2049 90

View Full Product Details