Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AIP1
Entrez Gene ID
855117
Gene Name
Gene Aliases
YMR092C
Chromosome Location
Chr.XIII: 451632 - 453479 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182592.1 NP_013810.1 1 - 1 1753 95

View Full Product Details