Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TIF11
Entrez Gene ID
855302
Gene Name
Gene Aliases
YMR260C
Chromosome Location
Chr.XIII: 789378 - 789839 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182767.1 NP_013987.1 1 - 1 354 102

View Full Product Details