Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MIC27
Entrez Gene ID
855623
Gene Name
Gene Aliases
AIM37, MCS27, YNL100W
Chromosome Location
Chr.XIV: 437613 - 438317 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182938.1 NP_014299.1 1 - 1 546 102

View Full Product Details