Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STB1
Entrez Gene ID
855407
Gene Name
Gene Aliases
YNL309W
Chromosome Location
Chr.XIV: 52662 - 53924 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183147.1 NP_014090.2 1 - 1 1152 80

View Full Product Details