Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSK2
Entrez Gene ID
854111
Gene Name
Gene Aliases
YOL045W
Chromosome Location
Chr.XV: 243497 - 246802 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183299.1 NP_014597.1 1 - 1 2050 93

View Full Product Details