Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ISW2
Entrez Gene ID
854480
Gene Name
Gene Aliases
YOR304W
Chromosome Location
Chr.XV: 884514 - 887876 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183724.1 NP_014948.1 1 - 1 3256 85

View Full Product Details