Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RRG7
Entrez Gene ID
854482
Gene Name
Gene Aliases
YOR305W
Chromosome Location
Chr.XV: 889022 - 889750 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183725.1 NP_014950.1 1 - 1 647 103

View Full Product Details