Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
INA17
Entrez Gene ID
856005
Gene Name
Gene Aliases
AIM43, FMP14, YPL099C
Chromosome Location
Chr.XVI: 363520 - 364068 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183913.1 NP_015226.1 1 - 1 346 114

View Full Product Details