Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOP53
Entrez Gene ID
855957
Gene Name
Gene Aliases
YPL146C
Chromosome Location
Chr.XVI: 276162 - 277529 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183960.1 NP_015179.1 1 - 1 1245 84

View Full Product Details