Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SUI3
Entrez Gene ID
855838
Gene Name
Gene Aliases
YPL237W
Chromosome Location
Chr.XVI: 100496 - 101353 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184051.1 NP_015087.1 1 - 1 445 109

View Full Product Details