Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TIE1
Entrez Gene ID
494022
Gene Name
tyrosine kinase with immunoglobulin like and EGF like domains 1
Gene Aliases
TIE-1
Chromosome Location
Chr.6: 167919662 - 167947253 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_021096882.1 XP_020952541.1 - 2828 95
XM_021096883.1 XP_020952542.1 - 9181 95
XM_021096884.1 XP_020952543.1 - 2737 95
XM_021096885.1 XP_020952544.1 - 9094 95
XM_021096886.1 XP_020952545.1 - 2551 95
XM_021096887.1 XP_020952546.1 - 2542 95
XM_021096888.1 XP_020952547.1 - 3158 95
XM_021096889.1 XP_020952548.1 - 9283 95
GenBank mRNA AJ867846.1 - 1 - 2 62 95

View Full Product Details