Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACNG1
Entrez Gene ID
733677
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 1
Gene Aliases
-
Chromosome Location
Chr.12: 13462589 - 13481651 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044605.1 NP_001038070.1 2 - 3 312 60
XM_021066261.1 XP_020921920.1 - 944 60
GenBank mRNA DQ323981.1 - 2 - 3 312 60

View Full Product Details