Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EFNA1
Entrez Gene ID
100144501
Gene Name
ephrin A1
Gene Aliases
-
Chromosome Location
Chr.4: 94666457 - 94673442 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123110.1 NP_001116582.1 3 - 4 455 82
XM_021088465.1 XP_020944124.1 - 494 82
GenBank mRNA DQ917646.1 - 3 - 4 455 82

View Full Product Details