Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP27B1
Entrez Gene ID
397055
Gene Name
25-hydroxyvitamin D3 1alpha-hydroxylase
Gene Aliases
-
Chromosome Location
Chr.5: 23053129 - 23058865 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213995.1 NP_999160.1 3 - 4 614 68
XM_021091124.1 XP_020946783.1 - 1009 68
GenBank mRNA AB018743.1 - 3 - 4 614 68

View Full Product Details