Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASIP
Entrez Gene ID
414439
Gene Name
agouti signaling protein
Gene Aliases
agouti
Chromosome Location
Chr.17: 37542615 - 37718787 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001648.4 NP_001001648.1 3 - 4 352 103
NM_001011646.1 NP_001011646.1 3 - 4 388 103
NM_001011647.1 NP_001011647.1 4 - 5 401 103
NM_001011648.1 NP_001011648.1 3 - 4 366 103
XM_005672859.3 XP_005672916.2 - 3269 103
XM_013985281.2 XP_013840735.2 - 1554 103
XM_021077312.1 XP_020932971.1 - 545 103
XM_021077313.1 XP_020932972.1 - 443 103
XM_021077314.1 XP_020932973.1 - 2094 103
GenBank mRNA AJ634674.1 - 3 - 4 352 103
AJ634675.1 - 3 - 4 388 103
AJ634676.2 - 4 - 5 401 103
AJ634677.1 - 3 - 4 366 103
AY308996.1 - 3 - 4 316 103
AY308997.1 - 3 - 4 316 103
AY308998.1 - 3 - 4 103
JQ839264.1 - 3 - 4 316 103

View Full Product Details