The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007194.3 | NP_001007195.1 | 4 - 5 | 417 | 126 |
GenBank mRNA | AY771222.2 | - | 4 - 5 | 417 | 126 |
EU421730.1 | - | 4 - 5 | 417 | 126 | |
HQ329180.1 | - | 3 - 4 | 376 | 126 | |
HQ329181.1 | - | 3 - 4 | 376 | 126 | |
HQ329182.1 | - | 3 - 4 | 376 | 126 |