Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSHZ2
Entrez Gene ID
100136903
Gene Name
teashirt zinc finger homeobox 2
Gene Aliases
-
Chromosome Location
Chr.17: 54111262 - 54374699 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114674.1 NP_001108146.1 2 - 2 3054 103
GenBank mRNA AK352199.1 - 2 - 2 3599 103
AK395280.1 - 2 - 2 3814 103

View Full Product Details