Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CASP10
Entrez Gene ID
100154896
Gene Name
caspase 10
Gene Aliases
-
Chromosome Location
Chr.15: 104858082 - 104895941 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161640.1 NP_001155112.1 7 - 8 1078 70
XM_021074526.1 XP_020930185.1 - 1097 70
GenBank mRNA AB499815.1 - 7 - 8 1078 70
AK232925.1 - 7 - 8 1034 70

View Full Product Details