Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPL52
Entrez Gene ID
100157971
Gene Name
mitochondrial ribosomal protein L52
Gene Aliases
-
Chromosome Location
Chr.7: 76187447 - 76191647 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001185151.1 NP_001172080.1 1 - 2 45 70
GenBank mRNA AK346805.1 - 1 - 2 45 70
BW963650.1 - 1 - 2 45 70

View Full Product Details