Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDIA6
Entrez Gene ID
100499564
Gene Name
protein disulfide isomerase family A member 6
Gene Aliases
pdi-p5
Chromosome Location
Chr.3: 125752079 - 125774225 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195119.1 NP_001182048.1 10 - 11 1159 91
GenBank mRNA AB282746.1 - 10 - 11 1159 91
AK234326.1 - 11 - 12 1207 91
AK238249.1 - 11 - 12 1207 91
AK350456.1 - 11 - 12 1204 91
AK391067.1 - 11 - 12 1236 91
AK400662.1 - 11 - 12 1260 91

View Full Product Details