The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142839.2 | NP_001136311.1 | 1 - 2 | 117 | 82 |
GenBank mRNA | AY609823.1 | - | 1 - 2 | 111 | 82 |
EU417836.1 | - | 1 - 2 | 117 | 82 |