Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDCBP2
Entrez Gene ID
100513633
Gene Name
syndecan binding protein 2
Gene Aliases
-
Chromosome Location
Chr.17: 33967075 - 33985975 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244863.1 NP_001231792.1 3 - 4 283 64
XM_013985196.2 XP_013840650.1 - 615 64
XM_021077266.1 XP_020932925.1 - 1280 64
XM_021077267.1 XP_020932926.1 - 560 64
GenBank mRNA AK349737.1 - 3 - 4 285 64

View Full Product Details