Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RTCA
Entrez Gene ID
100154102
Gene Name
RNA 3'-terminal phosphate cyclase
Gene Aliases
RTCD1
Chromosome Location
Chr.4: 117852776 - 117875630 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243470.1 NP_001230399.1 10 - 11 1085 97
GenBank mRNA AK235362.1 - 9 - 10 1039 97
AK346494.1 - 10 - 11 1085 97
AK394808.1 - 9 - 10 1108 97
AY610211.1 - - 952 97

View Full Product Details