Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INTS13
Entrez Gene ID
100155867
Gene Name
integrator complex subunit 13
Gene Aliases
ASUN, C5H12orf11
Chromosome Location
Chr.5: 46823746 - 46860552 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243595.1 NP_001230524.1 10 - 11 1270 86
XM_021090951.1 XP_020946610.1 - 1622 86
XM_021090952.1 XP_020946611.1 - 1233 86
XM_021090953.1 XP_020946612.1 - 1213 86
XM_021090954.1 XP_020946613.1 - 1219 86
GenBank mRNA AK343638.1 - 10 - 11 1270 86

View Full Product Details