Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRTO4
Entrez Gene ID
100511136
Gene Name
MRT4 homolog, ribosome maturation factor
Gene Aliases
-
Chromosome Location
Chr.6: 77747799 - 77753924 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243715.1 NP_001230644.1 3 - 4 235 63
GenBank mRNA AK240022.1 - 3 - 4 235 63
AK351341.1 - 3 - 4 235 63

View Full Product Details