Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRI
Entrez Gene ID
100514123
Gene Name
sorcin
Gene Aliases
-
Chromosome Location
Chr.9: 68492235 - 68515247 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244142.1 NP_001231071.1 2 - 3 157 76
NM_001244143.1 NP_001231072.1 2 - 3 149 76
GenBank mRNA AK231770.1 - 2 - 3 145 76
AK392529.1 - 2 - 3 142 76
AK395791.1 - 2 - 3 141 76
AK398866.1 - 2 - 3 141 76
AK399411.1 - 2 - 3 143 76
AK400616.1 - 2 - 3 141 76
AK400684.1 - 2 - 3 189 76
EW289129.2 - 2 - 3 133 76
EW326025.2 - 2 - 3 185 76
EW520500.2 - 2 - 3 189 76

View Full Product Details