Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEAK1
Entrez Gene ID
100152646
Gene Name
pseudopodium enriched atypical kinase 1
Gene Aliases
-
Chromosome Location
Chr.7: 57219506 - 57503807 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244188.1 NP_001231117.1 3 - 4 3411 104
XM_005656279.3 XP_005656336.1 - 5143 104
XM_021097992.1 XP_020953651.1 - 3821 104
XM_021097993.1 XP_020953652.1 - 3921 104
XM_021097994.1 XP_020953653.1 - 3658 104
XM_021097995.1 XP_020953654.1 - 3454 104

View Full Product Details