Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZCCHC9
Entrez Gene ID
100514154
Gene Name
zinc finger CCHC-type containing 9
Gene Aliases
-
Chromosome Location
Chr.2: 89840012 - 89849066 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244220.1 NP_001231149.1 3 - 4 648 77
GenBank mRNA BX915266.2 - 2 - 3 318 77

View Full Product Details