Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZCCHC9
Entrez Gene ID
100514154
Gene Name
zinc finger CCHC-type containing 9
Gene Aliases
-
Chromosome Location
Chr.2: 89840012 - 89849066 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244220.1 NP_001231149.1 5 - 6 812 80
GenBank mRNA AK346041.1 - 5 - 6 795 80
AK346392.1 - 5 - 6 812 80
BX915266.2 - 4 - 5 482 80

View Full Product Details