Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABHD14A
Entrez Gene ID
100625665
Gene Name
abhydrolase domain containing 14A
Gene Aliases
-
Chromosome Location
Chr.13: 34104359 - 34111953 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244243.2 NP_001231172.1 4 - 5 712 86
XM_005669556.3 XP_005669613.1 - 830 86
XM_005669557.3 XP_005669614.1 - 718 86
XM_005669558.3 XP_005669615.1 - 879 86
XM_021068476.1 XP_020924135.1 - 576 86
XM_021068477.1 XP_020924136.1 - 539 86
XM_021068478.1 XP_020924137.1 - 497 86
XM_021068479.1 XP_020924138.1 - 1243 86
XM_021068480.1 XP_020924139.1 - 846 86
XM_021068481.1 XP_020924140.1 - 835 86
XM_021068482.1 XP_020924141.1 - 835 86
XM_021068483.1 XP_020924142.1 - 794 86
XM_021068484.1 XP_020924143.1 - 745 86
XM_021068485.1 XP_020924144.1 - 644 86
XM_021068486.1 XP_020924145.1 - 650 86
XM_021068487.1 XP_020924146.1 - 2103 86
GenBank mRNA AK234347.1 - 4 - 5 713 86
AK391715.1 - 4 - 5 683 86

View Full Product Details