Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THUMPD1
Entrez Gene ID
100512402
Gene Name
THUMP domain containing 1
Gene Aliases
-
Chromosome Location
Chr.3: 25285682 - 25315710 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244282.1 NP_001231211.1 3 - 4 759 149
XM_005662087.3 XP_005662144.1 - 1093 149
XM_013995606.2 XP_013851060.1 - 1095 149
GenBank mRNA AK235689.1 - 3 - 4 747 149
AK395562.1 - 3 - 4 759 149
FS663292.1 - 1 - 2 170 149

View Full Product Details