Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOXRED2
Entrez Gene ID
100513010
Gene Name
FAD dependent oxidoreductase domain containing 2
Gene Aliases
-
Chromosome Location
Chr.5: 11233835 - 11254713 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244741.1 NP_001231670.1 1 - 2 1200 71
XM_005663826.3 XP_005663883.1 - 639 71

View Full Product Details