Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDC1
Entrez Gene ID
100232858
Gene Name
pyruvate decarboxylase 1
Gene Aliases
GSVIVT00001355001
Chromosome Location
Chr.8: 7060810 - 7063804 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF195868.1 - 1 - 2 438 61

View Full Product Details