Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DFR
Entrez Gene ID
100233141
Gene Name
dihydroflavonol reductase
Gene Aliases
dfr1
Chromosome Location
Chr.18: 10768605 - 10771673 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY780886.1 - 2 - 3 293 82
X75964.1 - 2 - 3 334 82
Y11749.1 - 2 - 3 338 82

View Full Product Details