Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TFL1C
Entrez Gene ID
100233010
Gene Name
TFL1C protein
Gene Aliases
GSVIVT00007370001
Chromosome Location
Chr.16_NW_002238120: 122710 - 123783 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA DQ871593.1 - 3 - 4 308 73

View Full Product Details