Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CBL03
Entrez Gene ID
100262221
Gene Name
calcineurin B-like protein 03
Gene Aliases
VITISV_029390
Chromosome Location
Chr.Un_NW_002240558: 24448 - 32544 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA FJ901269.1 - 2 - 3 212 89

View Full Product Details