Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ran
Entrez Gene ID
448091
Gene Name
RAN, member RAS oncogene family
Gene Aliases
RanTC4, ara24, gsp1, ran-1, tc4
Chromosome Location
Chr.1: 149220368 - 149228809 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004829.1 NP_001004829.1 2 - 3 283 69
XM_012959006.2 XP_012814460.1 2 - 3 85 69
XM_012959012.2 XP_012814466.1 2 - 3 233 69
XM_018091941.1 XP_017947430.1 2 - 3 184 69
GenBank mRNA BC074619.1 - 2 - 3 283 69
CR761555.2 - 1 - 2 48 69
CR761725.2 - 1 - 2 56 69

View Full Product Details