Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ran
Entrez Gene ID
448091
Gene Name
RAN, member RAS oncogene family
Gene Aliases
RanTC4, ara24, gsp1, ran-1, tc4
Chromosome Location
Chr.1: 149220368 - 149228809 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004829.1 NP_001004829.1 3 - 4 369 96
XM_012959006.2 XP_012814460.1 3 - 4 171 96
XM_012959012.2 XP_012814466.1 3 - 4 319 96
XM_018091941.1 XP_017947430.1 3 - 4 270 96
GenBank mRNA BC074619.1 - 3 - 4 369 96
CR761555.2 - 2 - 3 134 96
CR761725.2 - 2 - 3 142 96

View Full Product Details