Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ncs1
Entrez Gene ID
394570
Gene Name
neuronal calcium sensor 1
Gene Aliases
flup, freq, frequenin, ncs-1, xfreq
Chromosome Location
Chr.8: 5390295 - 5424766 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203642.1 NP_988973.1 1 - 2 409 96
GenBank mRNA BC061386.1 - 1 - 2 409 96
CR848567.2 - 1 - 2 201 96

View Full Product Details