Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ubqln1
Entrez Gene ID
394829
Gene Name
ubiquilin 1
Gene Aliases
da41, dsk2, plic-1, ubqln2, ubqn, xdrp1
Chromosome Location
Chr.1: 121848861 - 121859784 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203890.1 NP_989221.1 4 - 5 932 88
GenBank mRNA BC063190.1 - 4 - 5 932 88
CT025388.2 - 4 - 5 894 88

View Full Product Details