Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dhcr7
Entrez Gene ID
394844
Gene Name
7-dehydrocholesterol reductase
Gene Aliases
7-Dhcr, Xdhcr7, slos
Chromosome Location
Chr.Un|NW_016683485: 691 - 3141 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203904.1 NP_989235.1 - 449 65
GenBank mRNA BC063347.1 - - 449 65
CU075777.1 - - 425 65

View Full Product Details