Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
crp
Entrez Gene ID
448345
Gene Name
C-reactive protein, pentraxin-related
Gene Aliases
-
Chromosome Location
Chr.4: 117854577 - 117856649 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006707.1 NP_001006708.1 1 - 2 92 93
GenBank mRNA BC075426.1 - 1 - 2 92 93

View Full Product Details