Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fhl3
Entrez Gene ID
493527
Gene Name
four and a half LIM domains 3
Gene Aliases
slim2
Chromosome Location
Chr.2: 58142065 - 58195680 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008164.1 NP_001008165.1 4 - 5 633 92
XM_012956824.2 XP_012812278.1 5 - 6 783 92
XM_012956825.2 XP_012812279.1 4 - 5 543 92
GenBank mRNA BC082347.1 - 4 - 5 633 92
CR760328.2 - 4 - 5 633 92

View Full Product Details