Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
olig3
Entrez Gene ID
493553
Gene Name
oligodendrocyte transcription factor 3
Gene Aliases
bhlhb7
Chromosome Location
Chr.5: 64308105 - 64309932 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008190.1 NP_001008191.1 1 - 1 757 94
GenBank mRNA BC082516.1 - 1 - 1 757 94
CU075679.1 - 1 - 1 753 94

View Full Product Details