Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpf2
Entrez Gene ID
493256
Gene Name
ribosome production factor 2 homolog
Gene Aliases
bxdc1
Chromosome Location
Chr.5: 74086735 - 74100561 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008430.3 NP_001008430.2 8 - 9 683 93
XM_012962908.2 XP_012818362.1 8 - 9 867 93
XM_018093849.1 XP_017949338.1 6 - 7 734 93
GenBank mRNA BC080134.1 - 8 - 9 672 93
CF240668.1 - 8 - 9 672 93

View Full Product Details