Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tmed9
Entrez Gene ID
496564
Gene Name
transmembrane p24 trafficking protein 9
Gene Aliases
Xp24a2, Xp24alpha2
Chromosome Location
Chr.3: 38129612 - 38133482 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011147.1 NP_001011147.1 2 - 3 238 74
GenBank mRNA BC084535.1 - 2 - 3 238 74
CR760324.2 - 2 - 3 278 74

View Full Product Details