Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ppp4r2
Entrez Gene ID
496569
Gene Name
protein phosphatase 4 regulatory subunit 2
Gene Aliases
pp4r2, ppp4r2-a, ppp4r2-b
Chromosome Location
Chr.4: 129363559 - 129379022 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011151.1 NP_001011151.1 6 - 7 687 96
GenBank mRNA BC084901.1 - 6 - 7 687 96
CR760404.1 - 6 - 7 573 96

View Full Product Details